Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 9, 10 | 2362 | ||
XM_006712654.2 | XP_006712717.1 | 9, 10 | 2939 | |||
XM_006712655.3 | XP_006712718.1 | 5, 6 | 576 | |||
XM_011511530.2 | XP_011509832.1 | 8, 9 | 2074 | |||
XM_017004569.1 | XP_016860058.1 | 8, 9 | 2066 | |||
XM_017004570.1 | XP_016860059.1 | 8, 9 | 2066 | |||
XM_017004571.1 | XP_016860060.1 | 8, 9 | 2004 | |||
XM_017004572.1 | XP_016860061.1 | 6, 7 | 1551 | |||
GenBank mRNA | AB046783.2 | - | 8, 9 | 2083 | ||
AB053305.1 | - | 9, 10 | 2129 | |||
AF391100.1 | - | 9, 10 | 2129 | |||
AK291029.1 | - | 9, 10 | 2146 | |||
BC029174.1 | - | 9, 10 | 2132 | |||
BC071576.1 | - | 9, 10 | 2124 | 30347073 | ||
KU178735.1 | - | 8, 9 | 2006 | |||
KU178736.1 | - | 8, 9 | 1944 |