Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020944.2 | NP_065995.1 | 13 | 2512 | ||
XM_005251526.4 | XP_005251583.1 | 13 | 2491 | |||
XM_006716809.3 | XP_006716872.1 | 13 | 2557 | |||
XM_011517973.2 | XP_011516275.1 | 13 | 2117 | |||
XM_017014937.1 | XP_016870426.1 | 13 | 2473 | |||
XM_017014938.1 | XP_016870427.1 | 13 | 2557 | |||
XM_017014939.1 | XP_016870428.1 | 13 | 2051 | |||
XM_017014940.1 | XP_016870429.1 | 12 | 1898 | |||
XM_017014941.1 | XP_016870430.1 | 12 | 2320 | |||
XM_017014942.1 | XP_016870431.1 | 13 | 2130 | |||
XM_017014943.1 | XP_016870432.1 | 13 | 2112 | |||
XM_017014944.1 | XP_016870433.1 | 12 | 1575 | |||
XM_017014945.1 | XP_016870434.1 | 12 | 1557 | |||
XM_017014946.1 | XP_016870435.1 | 14 | 2168 | |||
GenBank mRNA | AB046825.1 | - | 13 | 2502 | ||
AF258662.1 | - | 10 | 1561 | |||
AJ309567.1 | - | 13 | 2512 | |||
AK027884.1 | - | 13 | 2175 | |||
AK057610.1 | - | 1 | 44 | |||
AK295967.1 | - | 14 | 2505 | |||
AK297435.1 | - | 11 | 1519 | |||
AL834306.1 | - | 13 | 2272 | |||
BC011363.1 | - | 13 | 2471 | 4339156 |