Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300985.1 | NP_001287914.1 | 13 | 1690 | ||
NM_001300986.1 | NP_001287915.1 | 13 | 1699 | |||
NM_001300987.1 | NP_001287916.1 | 14 | 1846 | |||
NM_001300988.1 | NP_001287917.1 | 13 | 1690 | |||
NM_001300989.1 | NP_001287918.1 | 13 | 1675 | |||
NM_001300990.1 | NP_001287919.1 | 12 | 1599 | |||
NM_021190.3 | NP_067013.1 | 13 | 1675 | |||
NR_125356.1 | 12 | 1533 | ||||
NR_125357.1 | 12 | 1641 | ||||
XM_011541874.2 | XP_011540176.1 | 13 | 1523 | |||
XM_011541875.2 | XP_011540177.1 | 13 | 1521 | |||
XM_011541876.2 | XP_011540178.1 | 13 | 1553 | |||
XM_017001951.1 | XP_016857440.1 | 12 | 1553 | |||
GenBank mRNA | AB051232.1 | - | 13 | 1497 | ||
AB051233.1 | - | 12 | 1407 | |||
AF176085.1 | - | 13 | 1467 | |||
AF530580.1 | - | 13 | 1485 | |||
AF530581.1 | - | 13 | 1485 | |||
AF530582.1 | - | 5 | 538 | |||
AF530583.1 | - | 5 | 553 | |||
AF530584.1 | - | 5 | 538 | |||
AF530585.1 | - | 5 | 553 | |||
AK022716.1 | - | 8 | 981 | |||
AK025333.1 | - | 3 | 338 | |||
AK092216.1 | - | 12 | 1539 | |||
AK295384.1 | - | 12 | 1454 | |||
AK299755.1 | - | 12 | 1284 | |||
AK299897.1 | - | 14 | 1627 | |||
AK299922.1 | - | 13 | 1513 | |||
BC016582.2 | - | 13 | 1444 | 3872881 |