Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130033.1 | NP_001123505.1 | 7 | 1243 | ||
NM_001130034.1 | NP_001123506.1 | 8 | 1089 | |||
NM_001130035.1 | NP_001123507.1 | 7 | 984 | |||
NM_001130036.1 | NP_001123508.1 | 6 | 1116 | |||
NM_021200.2 | NP_067023.1 | 8 | 1348 | |||
XM_011545192.2 | XP_011543494.1 | 8 | 1922 | |||
XM_011545193.2 | XP_011543495.1 | 7 | 1817 | |||
GenBank mRNA | AB209716.1 | - | 7 | 896 | ||
AF081583.1 | - | 7 | 865 | |||
AF093249.1 | - | 7 | 868 | |||
AF100612.1 | - | 8 | 973 | |||
AF101054.1 | - | 7 | 1239 | |||
AK054725.1 | - | 3 | 1001 | |||
AK289620.1 | - | 7 | 896 | |||
AK289909.1 | - | 7 | 862 | |||
AK296033.1 | - | 7 | 788 | |||
AK311158.1 | - | 8 | 741 | |||
BC008075.2 | - | 7 | 922 | 3050369 | ||
EB387039.1 | - | 1 | 499 | |||
U89715.1 | - | 8 | 1344 |