Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130033.1 | NP_001123505.1 | 7 | 1886 | ||
NM_001130034.1 | NP_001123506.1 | 8 | 1732 | |||
NM_001130035.1 | NP_001123507.1 | 7 | 1627 | |||
NM_001130036.1 | NP_001123508.1 | 6 | 1759 | |||
NM_021200.2 | NP_067023.1 | 8 | 1991 | |||
XM_011545192.2 | XP_011543494.1 | 8 | 2565 | |||
XM_011545193.2 | XP_011543495.1 | 7 | 2460 | |||
GenBank mRNA | AB209716.1 | - | 7 | 1539 | ||
AF081583.1 | - | 7 | 1509 | |||
AF093249.1 | - | 7 | 1511 | |||
AF100612.1 | - | 8 | 1616 | |||
AF101054.1 | - | 7 | 1882 | |||
AK054725.1 | - | 3 | 1644 | |||
AL712105.1 | - | 1 | 396 | |||
BC008075.2 | - | 7 | 1565 | 3050369 | ||
CK819503.1 | - | 1 | 166 | |||
U89715.1 | - | 8 | 1987 |