Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021215.3 | NP_067038.1 | 6 | 1107 | ||
XM_005260477.3 | XP_005260534.1 | 6 | 1191 | |||
XM_005260480.4 | XP_005260537.1 | 6 | 1328 | |||
XM_011528954.2 | XP_011527256.1 | 6 | 1191 | |||
XM_017027990.1 | XP_016883479.1 | 6 | 1191 | |||
XM_017027991.1 | XP_016883480.1 | 5 | 1057 | |||
GenBank mRNA | AK092567.1 | - | 1 | 62 | ||
AK312468.1 | - | 6 | 867 | |||
BC033629.1 | - | 6 | 891 | 5223673 | ||
DQ372938.1 | - | 6 | 980 |