Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA1 SIRNA Assays ›
Gene Symbol:
PLEKHA1
Gene Name:
pleckstrin homology domain containing A1
Gene Aliases:
TAPP1
Chromosome Location:
Chr.10: 122374578 - 122432355 on Build GRCh38
UniGene ID:
Hs.643512
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133394
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001974.2
NP_001001974.1
9, 10
1003
NM_001195608.1
NP_001182537.1
10, 11
963
NM_021622.4
NP_067635.2
9, 10
844
XM_005270016.1
XP_005270073.1
10, 11
1022
XM_005270019.1
XP_005270076.1
10, 11
1022
XM_005270020.1
XP_005270077.1
10, 11
1022
XM_005270021.4
XP_005270078.1
10, 11
1022
XM_011540017.1
XP_011538319.1
10, 11
1200
XM_011540018.1
XP_011538320.1
10, 11
1259
XM_011540019.1
XP_011538321.1
10, 11
911
XM_011540021.1
XP_011538323.1
9, 10
906
XM_011540022.1
XP_011538324.1
10, 11
1224
XM_011540024.1
XP_011538326.1
10, 11
907
XM_017016478.1
XP_016871967.1
10, 11
1022
XM_017016479.1
XP_016871968.1
10, 11
1022
XM_017016480.1
XP_016871969.1
10, 11
1259
XM_017016481.1
XP_016871970.1
10, 11
1200
XM_017016482.1
XP_016871971.1
10, 11
1224
XM_017016483.1
XP_016871972.1
10, 11
1259
XM_017016484.1
XP_016871973.1
10, 11
1200
XM_017016485.1
XP_016871974.1
9, 10
906
XM_017016486.1
XP_016871975.1
10, 11
1200
XM_017016487.1
XP_016871976.1
10, 11
1259
XM_017016488.1
XP_016871977.1
9, 10
906
XM_017016489.1
XP_016871978.1
10, 11
1022
XM_017016490.1
XP_016871979.1
10, 11
1259
XM_017016491.1
XP_016871980.1
9, 10
906
GenBank mRNA
AB209407.1
-
4, 5
2201
AF286160.1
-
9, 10
820
AK057463.1
-
10, 11
963
AK075159.1
-
3, 4
251
AK314660.1
-
9, 10
890
BC001136.2
-
9, 10
877
2988158
BC042458.1
-
10, 11
2218
4828008
BE785067.1
-
5, 6
513
BX647627.1
-
3, 4
7790
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
luteinization
spermatogenesis
androgen metabolic process
estrogen metabolic process
post-embryonic development
phosphatidylinositol 3-kinase signaling
ruffle organization
Leydig cell differentiation
multicellular organism growth
establishment of protein localization
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
B cell receptor signaling pathway
negative regulation of protein kinase B signaling
palate development
face morphogenesis
cellular response to hydrogen peroxide
protein binding
lipid binding
PDZ domain binding
phosphatidylinositol-3,4-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline