Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021629.3 | NP_067642.1 | 10 | 2475 | ||
XM_005247692.2 | XP_005247749.1 | 10 | 2396 | |||
XM_006713721.2 | XP_006713784.1 | 10 | 2406 | |||
GenBank mRNA | AK001890.1 | - | 10 | 2475 | ||
AK022599.1 | - | 1 | 1168 |