Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290185.1 | NP_001277114.1 | 5 | 1136 | ||
NM_021729.5 | NP_068375.3 | 5 | 921 | |||
XM_011542899.2 | XP_011541201.1 | 5 | 879 | |||
GenBank mRNA | AB027508.2 | - | 5 | 891 | ||
AF258558.1 | - | 3 | 422 | |||
AF308800.1 | - | 5 | 855 | |||
AK023279.1 | - | 5 | 892 | |||
AK056701.1 | - | 4 | 748 | |||
AK223158.1 | - | 5 | 881 | |||
AK302973.1 | - | 5 | 1068 | |||
AL834218.1 | - | 5 | 868 | |||
BC065563.1 | - | 5 | 855 | 6144541 |