Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098536.1 | NP_001092006.1 | 4 | 380 | ||
NM_003481.2 | NP_003472.2 | 4 | 374 | |||
GenBank mRNA | AK308021.1 | - | 4 | 364 | ||
BC004889.2 | - | 4 | 353 | 3936781 | ||
BC005139.2 | - | 4 | 351 | 3506801 | ||
CN276110.1 | - | 4 | 374 | |||
U35116.1 | - | 4 | 364 | |||
U47927.1 | - | 4 | 373 | |||
X91349.1 | - | 4 | 350 |