Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136039.2 | NP_001129511.1 | 3 | 571 | ||
NM_001142355.1 | NP_001135827.1 | 3 | 520 | |||
NM_001142356.1 | NP_001135828.1 | 2 | 506 | |||
NM_021824.3 | NP_068596.2 | 3 | 723 | |||
XM_005246742.3 | XP_005246799.1 | 3 | 786 | |||
XM_011511582.1 | XP_011509884.1 | 3 | 707 | |||
XM_011511583.1 | XP_011509885.1 | 3 | 1451 | |||
XM_011511584.1 | XP_011509886.1 | 3 | 1269 | |||
XM_011511585.1 | XP_011509887.1 | 3 | 1218 | |||
XM_017004630.1 | XP_016860119.1 | 3 | 968 | |||
GenBank mRNA | AB038949.1 | - | 3 | 684 | ||
AF060513.1 | - | 3 | 480 | |||
AF182416.1 | - | 3 | 645 | |||
AF283538.1 | - | 3 | 526 | |||
AK023378.1 | - | 3 | 669 | |||
AK225905.1 | - | 3 | 2046 | |||
AK314709.1 | - | 3 | 525 | |||
AL834430.1 | - | 2 | 346 | |||
AY251943.1 | - | 2 | 509 | |||
AY251944.1 | - | 2 | 509 | |||
BC007654.2 | - | 2 | 507 | 3346099 | ||
BP194872.1 | - | 3 | 520 |