Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 10 | 2746 | ||
NM_015482.1 | NP_056297.1 | 10 | 3205 | |||
NM_021945.6 | NP_068764.3 | 11 | 2991 | |||
NR_104448.1 | 12 | 2462 | ||||
XM_011514801.2 | XP_011513103.1 | 11 | 3292 | |||
XM_017011180.1 | XP_016866669.1 | 11 | 3769 | |||
XM_017011181.1 | XP_016866670.1 | 9 | 2518 | |||
XM_017011182.1 | XP_016866671.1 | 8 | 2800 | |||
XM_017011184.1 | XP_016866673.1 | 8 | 2455 | |||
XM_017011186.1 | XP_016866675.1 | 11 | 2688 | |||
GenBank mRNA | AK000691.1 | - | 1 | 1039 | ||
AK172770.1 | - | 7 | 2301 | |||
BC022217.1 | - | 1 | 368 | |||
BC090851.1 | - | 1 | 273 | |||
BC128580.1 | - | 10 | 2698 | 40128787 | ||
BC128581.1 | - | 12 | 2418 |