Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164831.2 | NP_001158303.1 | 8 | 1098 | ||
NM_001164832.2 | NP_001158304.1 | 9 | 1447 | |||
NM_001164836.2 | NP_001158308.1 | 10 | 1523 | |||
NM_001164837.2 | NP_001158309.1 | 9 | 1426 | |||
NM_022003.3 | NP_071286.1 | 8 | 1350 | |||
GenBank mRNA | AK055389.1 | - | 1 | 1289 | ||
AK092198.1 | - | 1 | 1840 | |||
AK290757.1 | - | 10 | 1223 | |||
AL136699.1 | - | 8 | 936 | |||
AL832811.1 | - | 9 | 1041 | |||
AY358976.1 | - | 8 | 957 | |||
BC018652.2 | - | 10 | 1131 | 2822864 | ||
BC093040.1 | - | 8 | 962 | 6526843 |