Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022048.3 | NP_071331.2 | 8 | 1317 | ||
GenBank mRNA | AB042562.1 | - | 7 | 965 | ||
AB042563.1 | - | 7 | 949 | |||
AF223357.2 | - | 7 | 1035 | |||
AK026957.1 | - | 4 | 424 | |||
AK225960.1 | - | 8 | 1247 | |||
AL832551.1 | - | 8 | 1095 | |||
BC017236.2 | - | 7 | 766 | |||
BC130613.1 | - | 8 | 1090 | 40146644 | ||
BC136504.1 | - | 8 | 1090 | 9020493 |