Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130455.1 | NP_001123927.1 | 6 | 712 | ||
NM_001130976.1 | NP_001124448.1 | 6 | 944 | |||
NM_001130977.1 | NP_001124449.1 | 6 | 944 | |||
NM_001130978.1 | NP_001124450.1 | 6 | 944 | |||
NM_001130979.1 | NP_001124451.1 | 7 | 1037 | |||
NM_001130980.1 | NP_001124452.1 | 7 | 1037 | |||
NM_001130981.1 | NP_001124453.1 | 7 | 1037 | |||
NM_001130982.1 | NP_001124454.1 | 7 | 805 | |||
NM_001130983.1 | NP_001124455.1 | 6 | 712 | |||
NM_001130984.1 | NP_001124456.1 | 6 | 712 | |||
NM_001130985.1 | NP_001124457.1 | 7 | 805 | |||
NM_001130986.1 | NP_001124458.1 | 6 | 712 | |||
NM_001130987.1 | NP_001124459.1 | 7 | 805 | |||
NM_003494.3 | NP_003485.1 | 6 | 944 | |||
XM_005264584.4 | XP_005264641.1 | 7 | 820 | |||
XM_005264585.4 | XP_005264642.1 | 7 | 1505 | |||
GenBank mRNA | AF075575.1 | - | 6 | 941 | ||
BC171803.1 | - | 6 | 917 | |||
DQ267935.1 | - | 6 | 712 | |||
EU515155.1 | - | 7 | 938 | |||
EU515156.1 | - | 6 | 845 | |||
EU515157.1 | - | 6 | 845 | |||
EU515158.1 | - | 7 | 938 | |||
EU515159.1 | - | 6 | 845 | |||
EU515160.1 | - | 7 | 938 | |||
EU515161.1 | - | 7 | 805 | |||
EU515162.1 | - | 6 | 712 | |||
EU515163.1 | - | 6 | 712 | |||
EU515164.1 | - | 7 | 805 | |||
EU515165.1 | - | 6 | 712 | |||
EU515166.1 | - | 7 | 805 |