Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022052.1 | NP_071335.1 | 19 | 1679 | ||
GenBank mRNA | AF346619.1 | - | 19 | 1679 | ||
AJ277527.1 | - | 19 | 1679 | |||
AK057086.1 | - | 8 | 2062 | |||
BC031616.1 | - | 19 | 1672 | 5170537 |