Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 4 | 341 | ||
NR_135218.1 | 4 | 341 | ||||
XM_006713285.1 | XP_006713348.1 | 4 | 499 | |||
XM_011533995.1 | XP_011532297.1 | 4 | 276 | |||
XM_017007018.1 | XP_016862507.1 | 4 | 303 | |||
GenBank mRNA | AK096353.1 | - | 4 | 255 | ||
AK126883.1 | - | 4 | 283 | |||
AK308130.1 | - | 4 | 283 | |||
AL832034.1 | - | 4 | 303 | |||
AL832138.1 | - | 4 | 278 | |||
AL832344.1 | - | 4 | 280 | |||
AY744152.1 | - | 3 | 224 | |||
BC130632.1 | - | 4 | 282 | 40146663 | ||
CR627051.1 | - | 4 | 276 | |||
DA334003.1 | - | 4 | 323 |