Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 36 | 3698 | ||
NR_135218.1 | 36 | 3938 | ||||
XM_006713285.1 | XP_006713348.1 | 36 | 3856 | |||
XM_011533995.1 | XP_011532297.1 | 36 | 3633 | |||
XM_017007018.1 | XP_016862507.1 | 36 | 3660 | |||
GenBank mRNA | AF370367.1 | - | 27 | 2852 | ||
AK022627.1 | - | 14 | 1524 | |||
AK026968.1 | - | 18 | 1952 | |||
AK090632.1 | - | 26 | 2740 | |||
AK096353.1 | - | 36 | 3612 | |||
AK126883.1 | - | 20 | 3405 | |||
AL136729.1 | - | 9 | 1082 | |||
AL832034.1 | - | 36 | 3660 | |||
AL832138.1 | - | 35 | 3949 | |||
AL832344.1 | - | 36 | 3636 | |||
AY744152.1 | - | 35 | 3581 | |||
BC041145.1 | - | 21 | 2231 | |||
BC057392.1 | - | 23 | 2388 | |||
BC088801.1 | - | 18 | 1959 | |||
BC130632.1 | - | 36 | 3639 | 40146663 | ||
CR627051.1 | - | 32 | 5270 |