Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003498.5 | NP_003489.1 | 2 | 997 | ||
XM_017023741.1 | XP_016879230.1 | 2 | 1025 | |||
XM_017023742.1 | XP_016879231.1 | 2 | 923 | |||
GenBank mRNA | AF070673.1 | - | 2 | 979 | ||
AK025795.1 | - | 1 | 69 | |||
AL161976.1 | - | 2 | 958 | |||
BC036100.1 | - | 2 | 975 | 5277179 | ||
BC036443.1 | - | 2 | 969 | 4823944 |