Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 6, 7 | 1088 | ||
XM_005260504.4 | XP_005260561.1 | 6, 7 | 1801 | |||
XM_011528979.2 | XP_011527281.1 | 5, 6 | 1456 | |||
XM_017028012.1 | XP_016883501.1 | 5, 6 | 859 | |||
GenBank mRNA | AF395833.1 | - | 6, 7 | 1065 | ||
AK092485.1 | - | 4, 5 | 1297 | |||
AK296989.1 | - | 3, 4 | 659 | |||
BC041331.2 | - | 3, 4 | 303 | |||
BC142981.1 | - | 3, 4 | 303 |