Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022104.3 | NP_071387.1 | 8, 9 | 1129 | ||
XM_011528980.2 | XP_011527282.1 | 8, 9 | 1180 | |||
XM_011528981.2 | XP_011527283.1 | 8, 9 | 1091 | |||
XM_017028013.1 | XP_016883502.1 | 8, 9 | 1290 | |||
XM_017028014.1 | XP_016883503.1 | 8, 9 | 1093 | |||
GenBank mRNA | AB050014.1 | - | 8, 9 | 1109 | ||
AK056553.1 | - | 1107 | ||||
AK299435.1 | - | 8, 9 | 1114 | |||
BC010005.2 | - | 8, 9 | 1101 | 4131938 | ||
BC013365.2 | - | 8, 9 | 1100 | 3640928 |