Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300913.1 | NP_001287842.1 | 3, 4 | 519 | ||
NM_022338.3 | NP_071733.1 | 3, 4 | 519 | |||
XM_005274053.3 | XP_005274110.1 | 3, 4 | 694 | |||
GenBank mRNA | AF264781.1 | - | 3, 4 | 479 | ||
AY358754.1 | - | 1, 2 | 81 | |||
BC011765.2 | - | 2, 3 | 368 | 3614861 | ||
BG701401.1 | - | 3, 4 | 524 | |||
BI862360.1 | - | 3, 4 | 493 | |||
BX384793.2 | - | 2, 3 | 321 |