Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 6 | 1790 | ||
NM_022720.6 | NP_073557.3 | 6 | 1790 | |||
XM_006724268.2 | XP_006724331.1 | 3 | 425 | |||
GenBank mRNA | AK025539.1 | - | 5 | 754 | ||
AK025780.1 | - | 1 | 66 | |||
AK222796.1 | - | 5 | 754 | |||
AY189282.1 | - | 2 | 84 | |||
BC009323.2 | - | 4 | 593 | |||
BC037564.1 | - | 6 | 1740 | |||
BC078147.1 | - | 5 | 1504 | 6014346 | ||
BX649187.1 | - | 3 | 682 | |||
CR456356.1 | - | 5 | 1448 |