Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024019.3 | NP_076924.1 | 2 | 1937 | ||
GenBank mRNA | BC036847.1 | - | 2 | 1937 | 5247719 |