Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020946.1 | NP_065997.1 | 20 | 1739 | ||
NM_024820.2 | NP_079096.2 | 20 | 1739 | |||
XM_005252109.2 | XP_005252166.1 | 21 | 1642 | |||
XM_005252111.4 | XP_005252168.1 | 20 | 2555 | |||
XM_005252113.4 | XP_005252170.1 | 20 | 2544 | |||
XM_006717195.3 | XP_006717258.1 | 20 | 2556 | |||
XM_011518882.2 | XP_011517184.1 | 21 | 2612 | |||
XM_011518883.2 | XP_011517185.1 | 20 | 2556 | |||
XM_011518884.2 | XP_011517186.1 | 20 | 2563 | |||
XM_011518885.2 | XP_011517187.1 | 20 | 2305 | |||
XM_011518886.2 | XP_011517188.1 | 21 | 2610 | |||
XM_011518887.2 | XP_011517189.1 | 21 | 2601 | |||
XM_017014947.1 | XP_016870436.1 | 20 | 2568 | |||
XM_017014948.1 | XP_016870437.1 | 20 | 1588 | |||
XM_017014949.1 | XP_016870438.1 | 20 | 2487 | |||
XM_017014950.1 | XP_016870439.1 | 19 | 2432 | |||
XM_017014951.1 | XP_016870440.1 | 19 | 1532 | |||
XM_017014952.1 | XP_016870441.1 | 19 | 1723 | |||
GenBank mRNA | AB046828.2 | - | 15 | 1369 | ||
AK024782.1 | - | 20 | 1727 | |||
AK074151.1 | - | 15 | 1167 | |||
AK225104.1 | - | 20 | 1727 | |||
AK295710.1 | - | 18 | 1600 | |||
AK299867.1 | - | 11 | 801 | |||
BC009616.2 | - | 19 | 1707 | |||
BC039703.1 | - | 20 | 1592 | 4999484 |