Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031271.3 | NP_112561.2 | 1 | 1396 | ||
XM_005273575.3 | XP_005273632.1 | 8 | 2764 | |||
XM_005273576.3 | XP_005273633.1 | 5 | 2326 | |||
XM_006716369.3 | XP_006716432.1 | 7 | 2656 | |||
XM_011544588.2 | XP_011542890.1 | 7 | 2620 | |||
XM_011544589.2 | XP_011542891.1 | 8 | 2729 | |||
XM_011544590.2 | XP_011542892.1 | 8 | 2995 | |||
XM_011544591.2 | XP_011542893.1 | 7 | 2657 | |||
XM_011544592.2 | XP_011542894.1 | 7 | 2595 | |||
XM_011544593.2 | XP_011542895.1 | 5 | 3552 | |||
XM_011544594.2 | XP_011542896.1 | 7 | 2620 | |||
GenBank mRNA | AF285605.1 | - | 1 | 1396 | ||
BC140934.1 | - | 1 | 1471 | 9056746 |