Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199867.1 | NP_001186796.1 | 17 | 3520 | ||
NM_031417.3 | NP_113605.2 | 18 | 3600 | |||
XM_006723307.3 | XP_006723370.1 | 16 | 3211 | |||
GenBank mRNA | AB049127.1 | - | 18 | 3566 | ||
AB058763.1 | - | 18 | 3274 | |||
AB073663.1 | - | 9 | 2349 | |||
AB088047.1 | - | 17 | 3486 | |||
AK027619.1 | - | 10 | 2531 | |||
BC071948.1 | - | 7 | 2063 |