Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153446.1 | NP_001146918.1 | 5 | 1534 | ||
NM_001153484.1 | NP_001146956.1 | 4 | 1627 | |||
NM_001153552.1 | NP_001147024.1 | 3 | 1543 | |||
NM_001153585.1 | NP_001147057.1 | 4 | 1667 | |||
NM_001153635.1 | NP_001147107.1 | 5 | 1531 | |||
NM_001153663.1 | NP_001147135.1 | 4 | 1624 | |||
NM_001153690.1 | NP_001147162.1 | 3 | 1583 | |||
NM_032547.2 | NP_115936.2 | 5 | 1632 | |||
GenBank mRNA | AF448857.1 | - | 3 | 1434 | ||
AK027797.1 | - | 5 | 1632 | |||
AK092794.1 | - | 1 | 2392 | |||
AL133580.1 | - | 2 | 1287 | |||
BC016511.1 | - | 4 | 1469 |