Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033051.3 | NP_149040.3 | 4 | 2410 | ||
GenBank mRNA | AK055571.1 | - | 5 | 2350 | ||
BC048285.1 | - | 4 | 2262 | 5213610 |