Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003551.2 | NP_003542.1 | 3 | 404 | ||
XM_005272099.1 | XP_005272156.1 | 2 | 331 | |||
XM_006714716.3 | XP_006714779.1 | 2 | 331 | |||
XM_017009945.1 | XP_016865434.1 | 2 | 331 | |||
GenBank mRNA | AF067724.1 | - | 3 | 374 | ||
AK312181.1 | - | 3 | 351 | |||
BC026182.2 | - | 3 | 376 | 5270050 | ||
KU178416.1 | - | 2 | 326 | |||
U90450.1 | - | 3 | 350 | |||
Y14992.1 | - | 2 | 340 |