Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054027.4 | NP_473368.1 | 1 | 407 | ||
XM_011514067.1 | XP_011512369.1 | 1 | 370 | |||
GenBank mRNA | AB046801.1 | - | 1 | 237 | ||
AF274753.1 | - | 1 | 144 | |||
AK222631.1 | - | 105 | ||||
AK315012.1 | - | 1 | 105 | |||
AL833238.1 | - | 1 | 229 | |||
AY358503.1 | - | 1 | 158 | |||
BC009835.2 | - | 1 | 332 | 3927236 | ||
BC014526.2 | - | 1 | 132 | 3837372 | ||
BU193060.1 | - | 1 | 399 | |||
DA126651.1 | - | 1 | 370 |