Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003561.2 | NP_003552.1 | 2 | 456 | ||
NR_133651.1 | 2 | 456 | ||||
XM_011522695.2 | XP_011520997.1 | 2 | 546 | |||
XM_011522696.2 | XP_011520998.1 | 2 | 546 | |||
XM_011522697.1 | XP_011520999.1 | 2 | 546 | |||
XM_011522698.1 | XP_011521000.1 | 2 | 546 | |||
GenBank mRNA | BC069539.1 | - | 2 | 296 | 7262248 | |
BC106731.1 | - | 2 | 456 | 40015660 | ||
BC106732.1 | - | 2 | 456 | 40015664 | ||
BC111804.1 | - | 2 | 456 | 40069347 | ||
BM739473.1 | - | 2 | 190 | |||
CR456885.1 | - | 2 | 112 | |||
U95301.1 | - | 2 | 552 |
Gene Symbol: LOC100652777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522789.1 | XP_011521091.1 | 2 | 546 | ||
XM_011522790.2 | XP_011521092.1 | 2 | 546 |