Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 5 | 617 | ||
NM_001007227.1 | NP_001007228.1 | 4 | 495 | |||
NM_001007228.1 | NP_001007229.1 | 3 | 443 | |||
NM_001007229.1 | NP_001007230.1 | 4 | 494 | |||
NM_001007230.1 | NP_001007231.1 | 5 | 546 | |||
NM_003563.3 | NP_003554.1 | 4 | 565 | |||
XM_005257723.4 | XP_005257780.1 | 4 | 789 | |||
XM_005257724.4 | XP_005257781.1 | 3 | 719 | |||
XM_017025204.1 | XP_016880693.1 | 2 | 232 | |||
GenBank mRNA | AJ000644.1 | - | 3 | 303 | ||
AK222589.1 | - | 3 | 268 | |||
AK293045.1 | - | 4 | 776 | |||
AK293587.1 | - | 3 | 385 | |||
AK300616.1 | - | 3 | 267 | |||
AK312691.1 | - | 3 | 279 | |||
AK316366.1 | - | 5 | 344 | |||
BC001269.1 | - | 4 | 449 | 3461565 | ||
BC003385.1 | - | 3 | 260 | 3445878 | ||
CB992730.1 | - | 5 | 518 |