Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300895.1 | NP_001287824.1 | 3, 4 | 678 | ||
NM_001300896.1 | NP_001287825.1 | 4, 5 | 562 | |||
NM_145200.3 | NP_660201.1 | 3, 4 | 610 | |||
XM_011545181.2 | XP_011543483.1 | 4, 5 | 615 | |||
XM_011545183.2 | XP_011543485.1 | 4, 5 | 414 | |||
XM_017018025.1 | XP_016873514.1 | 4, 5 | 900 | |||
GenBank mRNA | AY039217.1 | - | 3, 4 | 610 | ||
BC033167.1 | - | 4, 5 | 562 | 4549347 |