Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 13 | 2870 | ||
NM_152233.2 | NP_689419.2 | 14 | 2765 | |||
XM_006720224.1 | XP_006720287.1 | 13 | 2652 | |||
GenBank mRNA | AK124315.1 | - | 1 | 2061 | ||
BC001798.1 | - | 14 | 2807 | 3354467 |