Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297614.2 | NP_001284543.1 | 7 | 1038 | ||
NM_001304479.1 | NP_001291408.1 | 7 | 783 | |||
NM_020468.5 | NP_065201.1 | 5 | 906 | |||
NM_153816.5 | NP_722523.1 | 7 | 1038 | |||
XM_005248738.3 | XP_005248795.1 | 7 | 1011 | |||
XM_005248740.3 | XP_005248797.1 | 5 | 880 | |||
XM_011535977.2 | XP_011534279.1 | 6 | 454 | |||
XM_011535979.1 | XP_011534281.1 | 5 | 446 | |||
XM_017011090.1 | XP_016866579.1 | 7 | 520 | |||
XM_017011091.1 | XP_016866580.1 | 7 | 1011 | |||
XM_017011092.1 | XP_016866581.1 | 5 | 648 | |||
XM_017011093.1 | XP_016866582.1 | 7 | 725 | |||
XM_017011094.1 | XP_016866583.1 | 5 | 620 | |||
XM_017011096.1 | XP_016866585.1 | 7 | 753 | |||
XM_017011097.1 | XP_016866586.1 | 7 | 705 | |||
XM_017011099.1 | XP_016866588.1 | 5 | 621 | |||
XM_017011100.1 | XP_016866589.1 | 4 | 313 | |||
XM_017011101.1 | XP_016866590.1 | 6 | 454 | |||
GenBank mRNA | AK000362.1 | - | 7 | 727 | ||
AK026479.1 | - | 6 | 1170 | |||
AK095380.1 | - | 8 | 1237 | |||
AK125063.1 | - | 3 | 1090 | |||
AK295417.1 | - | 7 | 512 | |||
AY044865.1 | - | 5 | 447 | |||
BC005110.1 | - | 5 | 635 | 3959086 | ||
BC046520.2 | - | 7 | 493 | |||
BC068589.1 | - | 7 | 777 | 4818366 | ||
BC095419.1 | - | 7 | 623 | 5268442 |