Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNX14 SIRNA Assays ›
Gene Symbol:
SNX14
Gene Name:
sorting nexin 14
Gene Aliases:
RGS-PX2, SCAR20
Chromosome Location:
Chr.6: 85505496 - 85594156 on Build GRCh38
UniGene ID:
Hs.485871
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133715
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNX14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297614.2
NP_001284543.1
9
1246
NM_001304479.1
NP_001291408.1
9
991
NM_020468.5
NP_065201.1
7
1114
NM_153816.5
NP_722523.1
9
1246
XM_005248738.3
XP_005248795.1
9
1219
XM_005248740.3
XP_005248797.1
7
1088
XM_011535977.2
XP_011534279.1
8
662
XM_011535979.1
XP_011534281.1
7
654
XM_017011090.1
XP_016866579.1
9
728
XM_017011091.1
XP_016866580.1
9
1219
XM_017011092.1
XP_016866581.1
7
856
XM_017011093.1
XP_016866582.1
9
933
XM_017011094.1
XP_016866583.1
7
828
XM_017011095.1
XP_016866584.1
7
568
XM_017011096.1
XP_016866585.1
9
961
XM_017011097.1
XP_016866586.1
9
913
XM_017011098.1
XP_016866587.1
8
863
XM_017011099.1
XP_016866588.1
7
829
XM_017011100.1
XP_016866589.1
6
521
XM_017011101.1
XP_016866590.1
8
662
GenBank mRNA
AK000362.1
-
9
935
AK026479.1
-
8
1378
AK095380.1
-
10
1445
AK125063.1
-
5
1298
AK295417.1
-
9
720
AY044865.1
-
7
655
BC005110.1
-
7
843
3959086
BC046520.2
-
9
701
BC068589.1
-
9
985
4818366
BC095419.1
-
9
831
5268442
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
autophagosome maturation
phosphatidylinositol binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0