Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172231.3 | NP_757386.2 | 12 | 1671 | ||
XM_005260002.2 | XP_005260059.1 | 11 | 1686 | |||
XM_011528155.2 | XP_011526457.1 | 11 | 1564 | |||
GenBank mRNA | AF521128.1 | - | 12 | 1671 | ||
AK092745.1 | - | 4 | 1613 | |||
AK096152.1 | - | 11 | 1760 | |||
AK292085.1 | - | 12 | 1668 | |||
AL137286.1 | - | 6 | 959 | |||
AL713757.1 | - | 6 | 959 | |||
AY072916.1 | - | 12 | 1847 | |||
AY072917.1 | - | 12 | 1651 | |||
BC063784.1 | - | 12 | 1649 | |||
BC142988.1 | - | 12 | 1668 | 8860435 |