Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 2, 3 | 300 | ||
NM_177980.2 | NP_817089.1 | 14, 15 | 2403 | |||
XM_011528969.2 | XP_011527271.1 | 14, 15 | 2409 | |||
XM_011528970.2 | XP_011527272.1 | 8, 9 | 1425 | |||
XM_017027994.1 | XP_016883483.1 | 14, 15 | 3072 | |||
XM_017027996.1 | XP_016883485.1 | 14, 15 | 2974 | |||
XM_017027998.1 | XP_016883487.1 | 14, 15 | 3070 | |||
XM_017028000.1 | XP_016883489.1 | 13, 14 | 2242 | |||
XM_017028002.1 | XP_016883491.1 | 14, 15 | 3071 | |||
GenBank mRNA | AK055202.1 | - | 14, 15 | 2392 | ||
AK074477.1 | - | 2, 3 | 288 | |||
BC136306.1 | - | 14, 15 | 2207 | 9020293 | ||
BC143931.1 | - | 14, 15 | 2207 |