Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201457.1 | NP_001188386.1 | 12 | 1562 | ||
NM_031272.4 | NP_112562.3 | 12 | 1544 | |||
NM_198393.3 | NP_938207.2 | 12 | 1544 | |||
XM_011525029.2 | XP_011523331.1 | 12 | 1523 | |||
XM_017024860.1 | XP_016880349.1 | 10 | 1194 | |||
XM_017024861.1 | XP_016880350.1 | 11 | 1369 | |||
GenBank mRNA | AK128217.1 | - | 8 | 979 | ||
AK308299.1 | - | 12 | 1504 | |||
AL834143.1 | - | 12 | 1543 | |||
BC040526.1 | - | 12 | 1530 | 4831308 |