Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198795.1 | NP_942090.1 | 3, 4 | 548 | ||
XM_005269978.2 | XP_005270035.1 | 3, 4 | 529 | |||
XM_005269979.2 | XP_005270036.1 | 3, 4 | 562 | |||
XM_011539959.2 | XP_011538261.1 | 3, 4 | 901 | |||
XM_011539960.2 | XP_011538262.1 | 3, 4 | 491 | |||
XM_011539961.2 | XP_011538263.1 | 3, 4 | 473 | |||
XM_011539962.1 | XP_011538264.1 | 3, 4 | 855 | |||
XM_011539963.1 | XP_011538265.1 | 3, 4 | 562 | |||
XM_011539964.1 | XP_011538266.1 | 3, 4 | 562 | |||
XM_011539966.1 | XP_011538268.1 | 3, 4 | 562 | |||
XM_017016414.1 | XP_016871903.1 | 3, 4 | 562 | |||
XM_017016415.1 | XP_016871904.1 | 3, 4 | 562 | |||
GenBank mRNA | AK122916.1 | - | 3, 4 | 558 | ||
AL833340.1 | - | 3, 4 | 550 | |||
BC035010.2 | - | 3, 4 | 854 | |||
BC063133.1 | - | 3, 4 | 456 | 6710928 | ||
BX648664.1 | - | 3, 4 | 493 |