Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133764
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
15
1389
NM_001282212.1
NP_001269141.1
15
1615
NM_001282213.1
NP_001269142.1
16
1668
NM_001282214.1
NP_001269143.1
16
1656
NM_001282215.1
NP_001269144.1
16
1453
NM_001282216.1
NP_001269145.1
15
1404
NM_001320329.1
NP_001307258.1
16
2321
NM_016250.2
NP_057334.1
16
1511
NM_201535.1
NP_963293.1
17
1553
NM_201536.1
NP_963294.1
15
1444
NM_201537.1
NP_963831.1
16
1486
NM_201538.1
NP_963832.1
15
1421
NM_201539.1
NP_963833.1
16
1463
NM_201540.1
NP_963834.1
17
1530
NM_201541.1
NP_963835.1
16
1488
XM_011536996.2
XP_011535298.1
17
1540
XM_011536997.1
XP_011535299.1
17
1541
XM_011536998.1
XP_011535300.1
17
1531
XM_011536999.1
XP_011535301.1
15
1936
XM_011537001.1
XP_011535303.1
16
1494
XM_011537002.1
XP_011535304.1
16
1640
XM_017021480.1
XP_016876969.1
17
1642
XM_017021481.1
XP_016876970.1
17
1681
XM_017021482.1
XP_016876971.1
17
1690
XM_017021483.1
XP_016876972.1
16
1574
XM_017021484.1
XP_016876973.1
16
1616
XM_017021485.1
XP_016876974.1
17
1681
XM_017021486.1
XP_016876975.1
17
1691
XM_017021487.1
XP_016876976.1
16
1600
XM_017021488.1
XP_016876977.1
16
1641
XM_017021489.1
XP_016876978.1
16
1746
XM_017021490.1
XP_016876979.1
15
1538
XM_017021491.1
XP_016876980.1
15
1573
XM_017021492.1
XP_016876981.1
16
1644
XM_017021493.1
XP_016876982.1
16
1790
XM_017021494.1
XP_016876983.1
18
1546
XM_017021495.1
XP_016876984.1
17
1478
XM_017021496.1
XP_016876985.1
16
1477
XM_017021497.1
XP_016876986.1
11
1171
GenBank mRNA
AB033074.1
-
15
1373
AF159092.3
-
16
1434
AF304051.1
-
17
1553
AK024521.1
-
17
1488
AK057843.1
-
16
1457
AK057847.1
-
15
1421
AK090657.1
-
15
1360
AK096999.1
-
16
2321
AK098388.1
-
1481
AK130029.1
-
15
1359
AK293514.1
-
15
1389
AL136574.1
-
15
1398
BC010458.1
-
15
1392
4215141
BC011240.1
-
16
1413
4153602
BC013209.1
-
5
2157
BC093038.1
-
16
1487
4811718
BX247987.1
-
15
1474
BX248031.1
-
13
1263
BX647091.1
-
13
1586
BX647748.1
-
16
1603
CR749252.1
-
16
1656
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline