Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 18 | 4316 | ||
XM_005264237.3 | XP_005264294.1 | 19 | 4589 | |||
XM_005264238.3 | XP_005264295.1 | 18 | 3565 | |||
XM_017003741.1 | XP_016859230.1 | 20 | 4594 | |||
XM_017003742.1 | XP_016859231.1 | 19 | 3633 | |||
XM_017003743.1 | XP_016859232.1 | 16 | 3315 | |||
XM_017003744.1 | XP_016859233.1 | 15 | 3063 | |||
GenBank mRNA | AB015982.2 | - | 18 | 4329 | ||
BU173084.1 | - | 1 | 469 |