Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037174.1 | NP_001032251.1 | 6 | 995 | ||
NM_012097.3 | NP_036229.1 | 6 | 1251 | |||
NM_177985.2 | NP_817114.2 | 6 | 993 | |||
GenBank mRNA | AF087889.1 | - | 1181 | |||
AF100740.1 | - | 6 | 998 | |||
AK292714.1 | - | 6 | 1161 | |||
BC001254.1 | - | 6 | 1169 | 3453177 | ||
BM800611.1 | - | 1 | 372 |