Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012109.2 | NP_036241.1 | 8 | 1013 | ||
GenBank mRNA | AF186264.1 | - | 8 | 983 | ||
BC010446.1 | - | 8 | 954 | 4181442 |