Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012118.3 | NP_036250.2 | 2 | 446 | ||
GenBank mRNA | BC021963.2 | - | 2 | 409 | ||
BC023512.2 | - | 2 | 409 | |||
BC077723.1 | - | 2 | 409 | |||
BC113494.1 | - | 2 | 310 | 8322546 | ||
BC113500.1 | - | 2 | 310 | 8322552 | ||
HY067201.1 | - | 2 | 447 |