Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012118.3 | NP_036250.2 | 2 | 655 | ||
GenBank mRNA | AF183961.1 | - | 2 | 625 | ||
AF199493.1 | - | 1 | 108 | |||
BC021963.2 | - | 2 | 618 | |||
BC023512.2 | - | 2 | 618 | |||
BC077723.1 | - | 2 | 618 | |||
BC113494.1 | - | 2 | 519 | 8322546 | ||
BC113500.1 | - | 2 | 519 | 8322552 |