Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHORDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144073.1 | NP_001137545.1 | 3, 4 | 674 | ||
NM_012124.2 | NP_036256.2 | 4, 5 | 731 | |||
XM_017017541.1 | XP_016873030.1 | 4, 5 | 445 | |||
GenBank mRNA | AF123249.1 | - | 3, 4 | 369 | ||
AF192466.1 | - | 4, 5 | 405 | |||
AK225365.1 | - | 4, 5 | 433 | |||
AK225565.1 | - | 4, 5 | 416 | |||
AK290231.1 | - | 4, 5 | 731 | |||
AK312663.1 | - | 4, 5 | 416 | |||
BC072461.1 | - | 4, 5 | 380 | 6471725 |