Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193268.1 | NP_001180197.1 | 22 | 2661 | ||
NM_001193269.1 | NP_001180198.1 | 22 | 2565 | |||
NM_012155.2 | NP_036287.1 | 19 | 2026 | |||
NR_034098.1 | 17 | 1746 | ||||
XM_006723125.2 | XP_006723188.1 | 22 | 2742 | |||
XM_006723126.2 | XP_006723189.1 | 22 | 2550 | |||
XM_011526694.2 | XP_011524996.1 | 21 | 2649 | |||
XM_011526695.2 | XP_011524997.1 | 21 | 2610 | |||
XM_011526698.2 | XP_011525000.1 | 19 | 2068 | |||
XM_011526700.2 | XP_011525002.1 | 19 | 2014 | |||
XM_011526701.2 | XP_011525003.1 | 14 | 1617 | |||
XM_017026550.1 | XP_016882039.1 | 18 | 1910 | |||
GenBank mRNA | AB209773.1 | - | 16 | 1762 | ||
AF103939.1 | - | 19 | 2020 | |||
AK295905.1 | - | 22 | 2565 | |||
BC032630.1 | - | 17 | 1845 | 5402615 |