Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 14 | 1344 | ||
NM_012157.3 | NP_036289.3 | 15 | 1551 | |||
XM_005265014.2 | XP_005265071.1 | 15 | 1664 | |||
XM_005265015.3 | XP_005265072.1 | 15 | 1884 | |||
XM_005265016.2 | XP_005265073.1 | 15 | 1628 | |||
XM_005265017.3 | XP_005265074.1 | 15 | 1567 | |||
XM_011533559.2 | XP_011531861.1 | 16 | 1829 | |||
XM_017006092.1 | XP_016861581.1 | 16 | 1721 | |||
XM_017006093.1 | XP_016861582.1 | 15 | 1711 | |||
XM_017006094.1 | XP_016861583.1 | 16 | 1651 | |||
GenBank mRNA | AF176518.1 | - | 15 | 1486 | ||
AK001271.1 | - | 15 | 1551 | |||
AK001438.1 | - | 15 | 1519 | |||
AK096492.1 | - | 2 | 1966 | |||
AK297534.1 | - | 15 | 1535 | |||
AK303764.1 | - | 13 | 1411 | |||
AL049953.1 | - | 8 | 961 | |||
BC031556.1 | - | 15 | 1495 | 5197151 |